Learn More
BD Vacutainer Lithium Heparin Tubes
Plasma tube for clinical chemistry includes spray-dried lithium heparin as an anticoagulant.
283.00 CHF - 325.00 CHF
Spezifikation
Zusatzstoff | Lithium Heparin |
---|---|
Material | PET |
Verschlussfarbe | Green |
Markertyp | Paper |
Produktcode | Marke | Kapazität (metrisch) | Röhrchengröße | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Kapazität (metrisch) | Röhrchengröße | Preis | Menge & Verfügbarkeit | ||||
15753036
|
BD Vacutainer
368884 |
4 mL | 13 x 75 mm |
283.00 CHF
1000 Stück |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||
12834811
|
BD Vacutainer
368886 |
6 mL | 13 x 100 mm |
325.00 CHF
1000 Stück |
Verfügbar ab: 22-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||
Beschreibung
- Plasma tubes for clinical chemistry with spray-dried lithium heparin.
- Heparin acts as an anticoagulant as it develops an antithrombin complex.
- Plasma tubes should be gently inverted 180° and back 8-10 times.
- Correct mixing immediately after the blood sample has been taken is extremely important to avoid microclotting.
- The lithium heparin is spray dried onto the inner walls of the tubes using a special procedure so that the additive is evenly distributed to achieve the best possible solubility.
- For clinical chemistry, lithium heparin is generally preferred over sodium heparin.
Spezifikation
Lithium Heparin | |
PET | |
Green | |
Paper |