missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LILRA4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21153-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
ILT7 is a pDC-specific IgG-like transcript and is a member of the leukocyte immunoglobulin-like receptor gene family. This receptor, in complex with FcER1a, transduces ITAM-mediated signals which negatively modulate type I IFN production by pDC. ILT7 is thus a potential target to regulate Type 1 IFN production by pDC, which play a major role during viral infection, auto-immune diseases and anti-cancer immunity. Additionally, ILT7, along with CD123 and BDCA-2, can be used to identify or classify subsets of pDCalong with TLR expression and TLR mediated immune responses.Spezifikation
LILRA4 | |
Polyclonal | |
Unconjugated | |
LILRA4 | |
CD85g, ILT 7, ILT7, LILRA4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23547 | |
Antibody | |
IgG |
Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P59901 | |
LILRA4 | |
LILRA4 Fusion Protein Ag15721 | |
150 μL | |
Primary | |
Human, Mouse | |
Liquid |