missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LH, alpha Mouse, Clone: LHa/756, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 1081-MSM2-P1
Additional Details : Gewicht : 0.01000kg
Spezifikation
LH, alpha | |
Cytoplasmic, secreted | |
LHa/756 | |
Unconjugated | |
NP_000726 | |
Mouse | |
13kDa | |
Protein A/G Purified | |
RUO | |
1081 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
LHA | |
Recombinant full-length hLH alpha protein | |
Anterior Pituitary | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |