missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11355-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Anti-viral innate immune responses are triggered by pathogen-associated molecular patters (such as the accumulation of intracellular nucleic acids resulting from virus infections) and represent the first line of defense against numerous infectious organisms. The Toll-like receptors (TLR) 3, 7, 8, and 9 are expressed in immune cells and function as transmembrane pattern recognition receptors to detect foreign nucleic acids. Other proteins that play similar roles, such as RIG-1 and MDA5 are members of a CARD-helicase family and are expressed in the cytoplasm. A third protein, LGP2, is similar to RIG-1 and MDA5, except for lacking a homologous CARD domain. It is thought to act as an element of negative-feedback regulation of intracellular antiviral signaling. At least four isoforms of LGP2 are known to exist.Spezifikation
LGP2 | |
Polyclonal | |
Unconjugated | |
DHX58 | |
D11LGP2, D11lgp2e, DHX58, LGP2, Protein D11Lgp2 homolog, RIG I like receptor LGP2, RLR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
303538, 79132, 80861 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96C10, Q99J87 | |
Dhx58 | |
LGP2 Fusion Protein Ag1910 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |