missing translation for 'onlineSavingsMsg'
Learn More

LGP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

133.80 CHF - 381.34 CHF

Spezifikation

Antigen LGP2
Konzentration 0.17 mg/mL
Anwendungen Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Klassifikation Polyclonal
Konjugat Unconjugated
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16848373
Unterlagen ansehen Aktionsdetails
Proteintech
11355-1-AP-20UL
20 μL
133.80 CHF
20 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
16838373
Unterlagen ansehen Aktionsdetails
Proteintech
11355-1-AP-150UL
150 μL
381.34 CHF
150 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

Anti-viral innate immune responses are triggered by pathogen-associated molecular patters (such as the accumulation of intracellular nucleic acids resulting from virus infections) and represent the first line of defense against numerous infectious organisms. The Toll-like receptors (TLR) 3, 7, 8, and 9 are expressed in immune cells and function as transmembrane pattern recognition receptors to detect foreign nucleic acids. Other proteins that play similar roles, such as RIG-1 and MDA5 are members of a CARD-helicase family and are expressed in the cytoplasm. A third protein, LGP2, is similar to RIG-1 and MDA5, except for lacking a homologous CARD domain. It is thought to act as an element of negative-feedback regulation of intracellular antiviral signaling. At least four isoforms of LGP2 are known to exist.
Spezifikation

Spezifikation

LGP2
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
Q96C10, Q99J87
303538, 79132, 80861
LGP2 Fusion Protein Ag1910
Primary
-20°C
DHX58
0.17 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
D11LGP2, D11lgp2e, DHX58, LGP2, Protein D11Lgp2 homolog, RIG I like receptor LGP2, RLR
Dhx58
IgG
Antigen Affinity Chromatography
Antibody
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications