Learn More
LGI4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559874
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: DTLTHVDLRG NPFQCDCRVL WLLQWMPTVN ASVGTGACAG PASLSHMQLH HLDPKTFKCR AIELSWFQTV GESALSV Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.
The leucine-rich, glioma inactivated gene 4 (LGI4) is a member of the LGI family in which LGI1 is the exemplar. The LGI family consists of four of highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 has been identified as a candidate tumor suppressor gene for glioma and plays a role in autodominant lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predominant auditory symptoms. Despite its high homology with LGI1 and similar pattern of expression, mutations in LGI4 have not been found to be associated with ADTLE. However, the LGI4 gene is located in a region linked to benign familial infantile convulsions. Further study revealed that a GC-to-AT polymorphism was correlated with childhood absence epilepsy. Other studies showed that decreasing LGI4 expression in cultured cells inhibits myelination, indicating that LGI4 may play a role in neural development.Spezifikation
LGI4 | |
Polyclonal | |
Unconjugated | |
LGI4 | |
clp; leucine rich repeat LGI family member 4; leucine-rich glioma inactivated protein 4; leucine-rich glioma-inactivated gene 4; leucine-rich glioma-inactivated protein 4; Leucine-rich repeat LGI family member 4; leucine-rich repeat LGI family, member 4; LGI1-like protein 3; LGI4; Lgil3; UNQ6515/PRO21485 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
163175 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N135 | |
LGI4 | |
Recombinant protein corresponding to Human LGI4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only