Learn More
Thermo Scientific™ [Leu31, Pro34] Neuropeptid Y (1-36), Schwein
für zell- und molekularbiologische Anwendungen.
Marke: Thermo Scientific™ J66699.M¬
Additional Details : Gewicht : 0.00001kg
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
Porcine | |
-30°C to -10°C | |
4222.69 | |
1 mg |
HPLC | |
Fest | |
[Leu31, Pro34] Neuropeptide Y (1-36), porcine | |
RUO - research use only |
Nur für Forschungszwecke