Learn More
Uvex™ LED Head Torch and Adapter
Features two separate reflectors for distance and area lighting, with complete protection against dust and powerful water jets (IP 66).
15.50 CHF - 90.45 CHF
Produktcode | Marke | Beschreibung | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Beschreibung | Preis | Menge & Verfügbarkeit | |||||
17229201
|
Uvex™
9790064 |
90.45 CHF
1 Stück |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||||
17279201
|
Uvex™
9790069 |
Pheos Helmadapter |
15.50 CHF
1 Stück |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
- Magnetic adapters for easy mounting to the U-Cap Sport Bump Cap
- Suitable for combination with uvex pheos and uvex pheos alpine helmets when used with adapter 9790069
- Complete protection against dust and powerful water jets (IP 66)
- Depending on the light mode used, head torch can last for up to 150 hr.
- Luminous range from 3 to 80 meters, with a max flux of 100 lumens
- Operational in temperatures ranging from -20°C to 45°C
- Head torch includes a bump cap adapter and headband
- Requires 3 x AAA batteries (not included)