missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDLR Mouse anti-Human, PE, Clone: C7, BD Pharmingen™
Mouse Monoclonal Antibody
Marke: BD Biosciences 565653
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
LDLR Monoclonal antibody specifically detects LDLR in Cow , Human samples. It is validated for Flow cytometry.
Spezifikation
| LDLR | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Cell Biology | |
| Bovine, Human | |
| Purified |
| Flow Cytometry | |
| C7 (also known as IgG-C7) | |
| PE | |
| low-density lipoprotein receptor; LDL receptor; FH; FHC; LDLCQ2 | |
| Cow adrenal LDL receptor | |
| RUO | |
| Primary | |
| Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions and unconjugated antibody and free PE were removed. | |
| IgG2b κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Reseach Use Only.
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur