missing translation for 'onlineSavingsMsg'
Learn More

LASP1 Rabbit Recombinant Antibody, Proteintech

Product Code. p-200106576 Shop All Proteintech Products
Change view
Click to view available options
Menge:
20 μL
100 μL
Unit Size:
100 Mikroliter
20 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30235169 100 μL 100 Mikroliter
30233815 20 μL 20 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30235169 Supplier Proteintech Supplier No. 824911RR100UL

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Rabbit Recombinant Antibody

The LASP1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human LASP1. This antibody recognizes human, mouse, rat antigen. The LASP1 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.

Specifications

Antigen LASP1
Anwendungen Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA
Klassifikation Recombinant Monoclonal
Klon 4J2
Konzentration 250 μg/mL
Konjugat Unconjugated
Gensymbole Lasp-1; MLN50
Wirtsspezies Rabbit
Immunogen Recombinant Protein
Reinigungsverfahren Protein A purification
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 3927
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung Store at -20°C. Stable for one year after shipment.
Molekulargewicht des Antikörpers 35 kDa
Produkttyp Recombinant Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.