Learn More
LARS2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17097-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This gene encodes a class 1 aminoacyl-tRNA synthetase, mitochondrial leucyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid.Spezifikation
| LARS2 | |
| Polyclonal | |
| Unconjugated | |
| LARS2 | |
| KIAA0028, LARS2, Leucine tRNA ligase, LEURS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23395 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15031 | |
| LARS2 | |
| LARS2 Fusion Protein Ag10576 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |