missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthan-Standard: La @ 1000 μg/ml in 5% HNO3, VHG™
Description
Lanthan-Standard: La @ 1000 μg/ml in 5% HNO3
Specifications
Specifications
| Chemischer Name oder Material | Lanthanum |
| CAS | 7439-91-0 |
| Verfahren | ICP/ICP-MS |
| Bezeichnung der Analyten oder Bestandteile | Lanthanum |
| Versandbedingung | Room Temperature |
| Summenformel | La |
| Konzentration | 1000 μg/mL |
| Zertifizierung/Konformität | ISO 17034 |
| Synonym | La |
| InChI-Formel | InChI=1S/La |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?