missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP-2/CD107b Antibody (H4B4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 23 publications
235.00 CHF - 592.00 CHF
Spezifikation
| Antigen | LAMP-2/CD107b |
|---|---|
| Klon | H4B4 |
| Konzentration | 1.0 mg/mL |
| Verdünnung | Western Blot 1:1000, Simple Western 1:25, Flow Cytometry 1 ug per million cells, ELISA, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:10-1:100, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunohistochemistry-Frozen 1:100 - 1:200, Immunoblotting reported in scientific literature (PMID 29643474), CyTOF-ready, Knockout Validated |
| Anwendungen | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, CyTOF, Immunoassay |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18099405
|
Novus Biologicals
NBP2-22217SS |
0.025 mL |
235.00 CHF
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18720654
|
Novus Biologicals
NBP2-22217 |
592.00 CHF
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | ||||||
Beschreibung
LAMP-2/CD107b Monoclonal specifically detects LAMP-2/CD107b in Human, Mouse, Chicken, Rat (Negative) samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, CyTOF-ready, Knockout Validated.Spezifikation
| LAMP-2/CD107b | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, CyTOF, Immunoassay | |
| Unconjugated | |
| Mouse | |
| Cardiovascular Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cognition and Behavior, Growth and Development, Immunology, Innate Immunity, Lipid and Metabolism, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones | |
| P13473 | |
| 3920 | |
| LAMP-2/CD107b Antibody (H4B4) was made using a human adherent spleen cells. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 45 kDa |
| H4B4 | |
| Western Blot 1:1000, Simple Western 1:25, Flow Cytometry 1 ug per million cells, ELISA, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:10-1:100, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunohistochemistry-Frozen 1:100 - 1:200, Immunoblotting reported in scientific literature (PMID 29643474), CyTOF-ready, Knockout Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat (Negative) | |
| CD107 antigen-like family member B, CD107b, CD107b antigen, LAMP-2, LAMPB, LGP110, LGP-96, Lysosomal Associated Membrane Protein 2, lysosomal-associated membrane protein 2, lysosome-associated membrane glycoprotein 2, Lysosome-associated membrane protein | |
| LAMP2 | |
| IgG1 κ | |
| Protein G purified | |
| Human. Does not react with Mouse or Rat. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts