missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAG-3 (CD223) Mouse anti-Human, PE, Clone: T47-530, BD Pharmingen™
Mouse Monoclonal Antibody
211.00 CHF - 555.00 CHF
Spezifikation
Antigen | LAG-3 (CD223) |
---|---|
Klon | T47-530 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | PE |
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
15834429
|
BD Biosciences
565617 |
25 Tests |
211.00 CHF
1 Stück |
Verfügbar ab: 22-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15824429
|
BD Biosciences
565616 |
100 Tests |
555.00 CHF
1 Stück |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Description
LAG-3 (CD223) Monoclonal antibody specifically detects LAG-3 (CD223) in Human samples. It is validated for Flow cytometry.
LAG-3 (CD223) Monoclonal antibody specifically detects LAG-3 (CD223) in Human samples. It is validated for Flow cytometry.Specifications
LAG-3 (CD223) | |
Flow Cytometry | |
PE | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human LAG-3 Recombinant Protein | |
Primary |
T47-530 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
LAG3; CD223; FDC; Lymphocyte activation gene 3 protein; Protein FDC | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions and unconjugated antibody and free PE were removed. |
For Research Use Only.