missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAG-3 (CD223) Mouse anti-Human, PE, Clone: T47-530, BD Pharmingen™
Mouse Monoclonal Antibody
Marke: BD Biosciences 565617
Additional Details : Gewicht : 0.01000kg
Beschreibung
LAG-3 (CD223) Monoclonal antibody specifically detects LAG-3 (CD223) in Human samples. It is validated for Flow cytometry.
LAG-3 (CD223) Monoclonal antibody specifically detects LAG-3 (CD223) in Human samples. It is validated for Flow cytometry.Spezifikation
LAG-3 (CD223) | |
Monoclonal | |
PE | |
LAG3; CD223; FDC; Lymphocyte activation gene 3 protein; Protein FDC | |
Human LAG-3 Recombinant Protein | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions and unconjugated antibody and free PE were removed. | |
IgG1 κ |
Flow Cytometry | |
T47-530 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Cell Biology | |
Human | |
Purified |
For Research Use Only.