missing translation for 'onlineSavingsMsg'
Learn More

LACTB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 18195-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838254

  • 131.19 CHF / 20 Mikroliter

Verfügbar ab: 28-06-2022
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This antibody is specific to LACTB.

This gene encodes a protein from the large 39S subunit of the mitochondrial ribosome (mitoribosome). The encoded protein has some sequence similarity to prokaryotic beta-lactamases but most of the residues that are responsible for the beta-lactamase activity are not conserved between the two proteins.
Spezifikation

Spezifikation

LACTB
Polyclonal
Unconjugated
LACTB
G24, lactamase, beta, LACTB, MRPL56
Rabbit
Antigen Affinity Chromatography
RUO
114294, 300803, 80907
Antibody
IgG
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P83111, Q9EP89
Lactb
LACTB Fusion Protein Ag12233
20 μL
Primary
Human, Rat, Mouse
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote