Learn More
Invitrogen™ La Crosse Virus Monoclonal Antibody (8C2.2)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA110801
Beschreibung
MA1-10801 detects La Crosse Virus. MA1-10801 has been successfully used in ELISA, Western blot, immunofluorescence, immunoprecipitation, and viral neutralization applications. The MA1-10801 immunogen is La Crosse Virus grown on E6 Vero cells and purified by sucrose gradient centrifugation.
Spezifikation
| La Crosse Virus | |
| Monoclonal | |
| 0.9 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2b |
| ELISA, Immunoprecipitation, Neutralization, Western Blot, Immunocytochemistry | |
| 8C2.2 | |
| Unconjugated | |
| LCV | |
| La Crosse Virus grown on E6 Vero cells and purified by sucrose gradient centrifugation. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.