Learn More
L-Psicose, TRC
Specifications
Specifications
| Chemischer Name oder Material | L-Psicose |
| CAS | 16354-64-6 |
| Summenformel | C6H12O6 |
| Menge | 10 mg |
| Synonym | L-Ribo-2-hexulose |
| InChI-Formel | InChI=1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6+/m0/s1 |
| SMILES | OC[C@H](O)[C@H](O)[C@H](O)C(=O)CO |
| IUPAC-Name | (3S,4S,5S)-1,3,4,5,6-Pentahydroxyhexan-2-eins |
| Molekulargewicht (g/mol) | 180.16 |
| Formelmasse | 180.0634 |