missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Plastin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 13025-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
L-plastin is a leukocyte specific actin bundling protein from fimbrin family of proteins. It has two actin-binding domains and two EF hand type calcium-binding domains. It gets phosphorylated on the serine residue in its headpiece region in response to a variety of inflammatory stimuli. It is hypothesized that L-plastin may play a role in regulating integrin mediated adhesion in leukocytes. L-plastin is expressed in majority of human cancer cell lines derived from solid tumors. Its expression is positively upregulated by testosteronse in andrgen receptor positive prostrate and breast cancer cells through three androgen receptor binding elements located in the upstream region of the gene.Spezifikation
L-Plastin | |
Polyclonal | |
Unconjugated | |
LCP1 | |
L PLASTIN, LCP 1, LCP1, Lymphocyte cytosolic protein 1, Plastin 2, PLS2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18826, 306071, 3936 | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P13796, Q61233 | |
Lcp1 | |
L-Plastin Fusion Protein Ag3571 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Liquid |