Learn More
Honeywell Fluka 1LT L-(+)-Lactic acid solution volumetric, 1 M C3H6O3 (1N)
Marke: Honeywell Fluka 35202-1L
1LT L-(+)-Lactic acid solution volumetric, 1 M C3H6O3 (1N)
Additional Details : CAS : 79-33-4 Gewicht : 1.03000kg
Marke: Honeywell Fluka 35202-1L
1LT L-(+)-Lactic acid solution volumetric, 1 M C3H6O3 (1N)
Additional Details : CAS : 79-33-4 Gewicht : 1.03000kg