missing translation for 'onlineSavingsMsg'
Learn More

L-Milchsäure, 90 %, TRC

Product Code. 30135744
Change view
Click to view available options
Menge:
1 g
5 g
Unit Size:
1 Gramm
5 Gramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30135744 5 g 5 Gramm
17991155 1 g 1 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135744 Supplier Toronto Research Chemicals Supplier No. TRCL1135005G
 Eingeschränkter Artikel

Dieser Artikel erfordert eine Preisaktualisierung, bevor er bestellt werden kann.

Fordern Sie hier einen Preis für diesen Artikel an.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-33-4
Summenformel C3 H6 O3
Molekulargewicht (g/mol) 90.08
Synonym S-Lactic acid, (S)-2-hydroxypropanoic acid
IUPAC-Name (2S)-2-Hydroxypropionsäure
SMILES C[C@H](O)C(=O)O

Specifications

Chemischer Name oder Material L-(+)-Lactic acid
InChI-Formel InChI=1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m0/s1
Formelmasse 90.0317
Menge 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.