missing translation for 'onlineSavingsMsg'
Learn More

L-Glutaminsäure-dibenzylester-p-toluolsulfonat, 98 %, Thermo Scientific™

Catalog Number 15457746
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2791-84-6
C26H29NO7S
499.578
MFCD00038980
HVZUAIVKRYGQRM-LMOVPXPDSA-N
h-glu obzl-obzl.tosoh, l-glutamic acid dibenzyl ester tosylate, s-dibenzyl 2-aminopentanedioate 4-methylbenzenesulfonate, h-glu obzl-obzl p-tosylate, l-glutamic acid dibenzyl ester 4-toluenesulfonate, glu obzl-obzl.tosoh, dibenzyl l-glutamate p-toluenesulfonate, h-glu obzl-obzl 4-tosylate, l-glutamic acid dibenzyl ester p-toluenesulfonate, h-glu obzl-obzl inverted exclamation mark currencytosoh
13233366
Dibenzyl (2S)-2-aminopentanedioat;4-Methylbenzolsulfonsäure
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CCC(C(=O)OCC2=CC=CC=C2)N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2791-84-6
C26H29NO7S
499.578
MFCD00038980
HVZUAIVKRYGQRM-LMOVPXPDSA-N
h-glu obzl-obzl.tosoh, l-glutamic acid dibenzyl ester tosylate, s-dibenzyl 2-aminopentanedioate 4-methylbenzenesulfonate, h-glu obzl-obzl p-tosylate, l-glutamic acid dibenzyl ester 4-toluenesulfonate, glu obzl-obzl.tosoh, dibenzyl l-glutamate p-toluenesulfonate, h-glu obzl-obzl 4-tosylate, l-glutamic acid dibenzyl ester p-toluenesulfonate, h-glu obzl-obzl inverted exclamation mark currencytosoh
13233366
Dibenzyl (2S)-2-aminopentanedioat;4-Methylbenzolsulfonsäure
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CCC(C(=O)OCC2=CC=CC=C2)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material L-Glutamic acid dibenzyl ester p-toluenesulfonate
Formelmasse 499.58
Reinheit (%) 98%
Menge 1 g
Physikalische Form Pulver
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'