missing translation for 'onlineSavingsMsg'
Learn More

L-Cystin, Acros Organics™

Marke:  Acros Organics 406160010

Additional Details : CAS : 56-89-3 Gewicht : 1.00000kg

Menge 1kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15279916

  • 505.60 CHF / 1 Kilogramm


Verfügbar ab
17-08-2020
Zum Warenkorb hinzufügen

Spezifikation

Chemische Identifikatoren

56-89-3
240.292
l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
CHEBI:16283
C(C(C(=O)O)N)SSCC(C(=O)O)N
C6H12N2O4S2
LEVWYRKDKASIDU-IMJSIDKUSA-N
67678
(2R)-2-amino-3[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propansäure

Spezifikation

L-Cystine
56-89-3
C6H12N2O4S2
Authentic
0.5% max. (1 g, 105°C)
Solubility in water: 0.112g/l(25°C), 24mg/100mL(50°C). Other solubilities: 52 mg/100mL water (75°C),114 mg/100mL water(100°C), soluble in aq. solutions <ph 2, soluble in mineral acids, alkalis, ammonium hydrox, ide,, insoluble in ethanol, diethyl ether, b
C(C(C(=O)O)N)SSCC(C(=O)O)N
240.292
CHEBI:16283
−224.00
300ppm max.
−224.00 (20.00°C c=2;1 M HCL dry.sub.)
1kg
200ppm max.
10ppm max.
[-SCH2CH(NH2)CO2H]2
10ppm max.
l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
LEVWYRKDKASIDU-IMJSIDKUSA-N
(2R)-2-amino-3[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propansäure
67678
240.3
Solubility in water: 0.112g/l(25°C), 24mg/100mL(50°C). Other solubilities: 52 mg/100mL water (75°C),114 mg/100mL water(100°C), soluble in aq. solutions <ph 2, soluble in mineral acids, alkalis, ammonium hydrox, ide,, insoluble in ethanol, diethyl ether, b
0.1% max. (1 g)
260.0°C to 261.0°C
Videos
Dokumentation