Learn More
L-Cysteinylglycine, TRC
Artikelnummer. 30134646
Ansicht ändern
Click to view available options
Menge:
25 mg
50 mg
Packungsgröße:
25 Milligramm
50 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30134646 25 mg 25 Milligramm
30008879 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134646 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCC99477025MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 19246-18-5
Synonym N-L-Cysteinylglycine∣144: PN: US20130123467 SEQID: 173 Claimed Protein∣145: PN: WO2012013136 SEQID: 39 Unclaimed Protein∣27: PN: WO2011133608 TABLE: 5 Claimed Sequence∣52: PN: EP2161028 PAGE: 10 Claimed Protein∣6: PN: WO2011003622 SEQID: 6 Claimed Protein∣Cysteinylglycine∣L-Cysteinylglycine;
Spezifikation
Reinheit >95% (HPLC)
Format Neat
Summenformel C5H10N2O3S
MDL-Nummer MFCD00055567
Molekulargewicht 178.21
InChI-Schlüssel ZUKPVRWZDMRIEO-VKHMYHEASA-N
SMILES N[C@@H](CS)C(=O)NCC(O)=O
IUPAC-Name 2-[(2R)-2-amino-3-sulfanylpropanamido]acetic acid
Produkttyp Other
Menge 25 mg
Molekulargewicht (g/mol) 178.21
Versandbedingung Room Temperature