missing translation for 'onlineSavingsMsg'
Learn More

L-Alanyl-Diphenylborinat, 95 %, Thermo Scientific™

Catalog Number p-7036844
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
116628-50-3
C15H16BNO2
253.108
MFCD22125132
BKYIQCXZZVWSLF-LBPRGKRZSA-N
diphenylboranyl 2s-2-aminopropanoate, l-alanyl diphenylborinate
99738129
Diphenylboranyl (2S)-2-aminopropanoat
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(C)N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
116628-50-3
C15H16BNO2
253.108
MFCD22125132
BKYIQCXZZVWSLF-LBPRGKRZSA-N
diphenylboranyl 2s-2-aminopropanoate, l-alanyl diphenylborinate
99738129
Diphenylboranyl (2S)-2-aminopropanoat
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(C)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material L-Alanyl diphenylborinate
Menge 250 mg
Formelmasse 253.11
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'