Learn More
KU-55933, MedChemExpress
Artikelnummer. 16464918
Encompass
Ansicht ändern
Click to view available options
Menge:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
16464918 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 16464918 Lieferant Medchem Express Lieferanten-Nr. HY1201610mg
 Eingeschränkter Artikel
Verfügbar ab: 30-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
MedChemExpress KU-55933 is a potent ATM inhibitor with an IC50 and Ki of 12.9 and 2.2 nM, respectively, and is highly selective for ATM as compared to DNA-PK, PI3K/PI4K, ATR and mTOR.
KU-55933 is a potent ATM inhibitor with an IC50 and Ki of 12.9 and 2.2 nM, respectively, and is highly selective for ATM as compared to DNA-PK, PI3K/PI4K, ATR and mTOR.
Spezifikation
Chemischer Name oder Material KU-55933
Physikalische Form Solid
CAS 587871-26-9
Farbe Earth Yellow
Menge 10 mg
Hinweise zur Reinheitsqualität Research
Summenformel C21H17NO3S2
Zur Verwendung mit (Anwendung) Cancer-Kinase/protease
Löslichkeitsinformationen DMSO : 80 mg/mL (202.28 mM; Need ultrasonic)
SMILES O=C1C=C(OC(C2=C3SC4=C(SC3=CC=C2)C=CC=C4)=C1)N5CCOCC5
Molekulargewicht (g/mol) 395.49
Formelmasse 395.49
Reinheit (%) 99.72%
Güte Research