missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT75 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA567414
Additional Details : Gewicht : 0.01000kg
Beschreibung
Immunogen sequence: LYNLGGAKRVS INGCGSSCRS GFGGRASNRF GVNSGFGYGG GVGGGF Highest antigen sequence identity to the following orthologs - mouse 71%, rat 60%.
KRT75 Polyclonal antibody specifically detects KRT75 in Human samples. It is validated for Immunocytochemistry.Spezifikation
KRT75 | |
Polyclonal | |
Unconjugated | |
KRT75 | |
4732468K03Rik; AA589387; CK-75; cytokeratin KRT2-6HF; cytokeratin type II; cytokeratin-75; hK6hf; K6HF; K75; KB18; keratin 75; keratin 75, type II; keratin, type II cytoskeletal 75; Keratin-6 hair follicle; keratin-75; keratinocyte associated protein 1; Krt2-6hf; KRT75; Krtcap1; mK6hf; PFB; type II keratin Kb18; type II keratin-18; type II keratin-K6hf; Type-II keratin Kb18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9119 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95678 | |
KRT75 | |
Recombinant Human KRT75 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only