missing translation for 'onlineSavingsMsg'
Learn More

KLRG1 Monoclonal Antibody (13F12F2), Super Bright™ 702, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Affymetrix eBioscience 67-9488-42

453.15 CHF gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21653" um Ihren Promotions-Preis zu erhalten.



Additional Details : Gewicht : 1.00000kg

Artikelnummer. 15824202

  • 477.00 CHF / 100 Stück
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Description: This 13F12F2 monoclonal antibody reacts with human killer cell lectin-like receptor subfamily G, member 1 (KLRG1), a type II transmembrane inhibitor receptor of the C-type lectin superfamily that contains an ITIM domain. This inhibitory receptor is expressed on subsets of gamma-delta T cells, NK (CD56dim), CD8+ and CD4+ T Cells. KLRG1 is expressed primarily by cells with an effector/memory phenotype that are short-lived, but capable of immediate effector cell function. Cadherin-E, -N, and -R are ligands for KLRG1. Cadherin/KLRG1 interaction inhibits cytolytic activity and proliferation. The percentage of KLRG1 positive cells can vary considerably, depending on antigen experience. The clones 13F12F2 and 13A2 appear to recognize a similar epitope based on cross-blocking studies. Applications Reported: This 13F12F2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 13F12F2 antibody has been pre-titrated and tested by flow cytometric analysis of human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 702 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 702 nm. We recommend using a 710/50 bandpass filter.

KLRG1 Monoclonal antibody specifically detects KLRG1 in Human samples. It is validated for Flow Cytometry.
Spezifikation

Spezifikation

KLRG1
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
Q96E93
Klrg1
Affinity chromatography
RUO
10219
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
13F12F2
Super Bright 702
Klrg1
2F1; 2F1-Ag; CLEC15A; C-type lectin domain family 15 member A; C-type lectin domain family 15, member A; ITIM-containing receptor MAFA-L; killer cell lectin like receptor G1; killer cell lectin-like receptor subfamily G member 1; killer cell lectin-like receptor subfamily G, member 1; Klrg1; Mafa; MAFA-2F1; MAFAL; MAFA-L; MAFA-LIKE; MAFA-like receptor; Mast cell function-associated antigen; mast cell function-associated antigen (ITIM-containing); mast cell function-associated antigen 2F1; natural killer cell receptor
Mouse
100 Tests
Primary
Human
Antibody
IgG2a κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only