missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRG1 Hamster anti-Mouse, BV786, Clone: 2F1, BD Horizon™
Hamster Monoclonal Antibody
Marke: BD Biosciences 565477
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
KLRG1 Monoclonal antibody specifically detects KLRG1 in Mouse samples. It is validated for Flow cytometry.
Spezifikation
| KLRG1 | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Hamster | |
| 50 μg | |
| Cell Biology | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 2F1 | |
| Brilliant Violet 786 | |
| Klrg1; Killer cell lectin-like receptor subfamily G member 1; MAFA; 2F1 Ag | |
| A-LAK from C57BL/6 mice | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV786 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV786 were removed. | |
| IgG2 κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Reseach Use Only.
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur