missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21191-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by TCP1 epsilon is a molecular chaperone that is a member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Mutations in TCP1 epsilon cause hereditary sensory and autonomic neuropathy with spastic paraplegia (HSNSP). Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 5 and 13.Spezifikation
KLHL4 | |
Polyclonal | |
Unconjugated | |
KLHL4 | |
DKELCHL, kelch like 4 (Drosophila), Kelch like protein 4, KHL4, KIAA1687, KLHL4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
237010, 317196, 56062 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9C0H6 | |
Klhl4 | |
KLHL4 Fusion Protein Ag15529 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |