missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIT [A829P] Recombinant Human Protein
Human KIT, C-terminal fragment, amino acids T544-V976 (as in NCBI/Protein entry NP_000213.1) with an A829P mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells.
574.00 CHF - 8710.00 CHF
Spezifikation
Zur Verwendung mit (Anwendung) | Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation |
---|---|
Zusammensetzung | 50mM HEPES pH 7.5, 100mM NaCl, 5mM DTT, 15mM reduced glutathione, 20% glycerol |
Molekulargewicht | 77.5kDa |
Lagerungsbedingungen | Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. |
Gen-Alias | CD117, PBT, SCFR, c-Kit |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15478863
|
Invitrogen™
PV6193 |
10μg |
574.00 CHF
10 Mikrogramm |
Verfügbar ab: 02-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15488863
|
Invitrogen™
PV6194 |
100μg |
1942.00 CHF
100 Mikrogramm |
Verfügbar ab: 02-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15498863
|
Invitrogen™
PV6195 |
1mg |
8710.00 CHF
1 Milligramm |
Verfügbar ab: 02-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Spezifikation
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
77.5kDa | |
CD117, PBT, SCFR, c-Kit | |
Human | |
GST Tag, His Tag |
50mM HEPES pH 7.5, 100mM NaCl, 5mM DTT, 15mM reduced glutathione, 20% glycerol | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
aa 544-976 | |
Recombinant |