missing translation for 'onlineSavingsMsg'
Learn More

Kindlin Recombinant Rabbit Monoclonal Antibody (7H7L9), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Marke:  Invitrogen 701830

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 15682015

  • 610.00 CHF / 100 Mikrogramm
Verfügbar ab: 10-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This antibody is predicted to react with Monkey, Equine, Bovine, Pig and Rabbit. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Kindlin Recombinant Monoclonal antibody specifically detects Kindlin in Human samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

Kindlin
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q9BQL6
FERMT1
Protein corresponding to Human FERMT1 (aa 200-409)
100 μg
Primary
Human
Antibody
IgG
Immunocytochemistry, Western Blot
7H7L9
Unconjugated
FERMT1
5830467P10Rik; C20orf42; DTGCU2; Fermitin family homolog 1; fermitin family homolog 1 (Drosophila); fermitin family member 1; FERMT1; FLJ20116; FLJ23423; KIND1; Kindlerin; kindlin 1; kindlin syndrome protein; Kindlin-1; RGD1306816; RP5-1056H1.1; UNC112 related protein 1; UNC-112 related protein 1; UNC112A; unc-112-related protein 1; URP1
Rabbit
Protein A
RUO
55612
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote