missing translation for 'onlineSavingsMsg'
Learn More

Kindlin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 11453-1-AP-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838543

  • 351.68 CHF / 150 Mikroliter

Verfügbar ab: 28-06-2022
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

11453-1-AP is a rabbit polyclonal antibody against human FERMT2.

This gene encodes a myosin IIA heavy chain that contains an IQ domain and a myosin head-like domain. The protein is involved in several important functions, including cytokinesis, cell motility and maintece of cell shape. Defects in MYH9 are the cause of non-syndromic sensorineural deafness autosomal domit type 17, Epstein syndrome, Alport syndrome with macrothrombocytopenia, Sebastian syndrome, Fechtner syndrome and macrothrombocytopenia with progressive sensorineural deafness.
Spezifikation

Spezifikation

Kindlin 2
Polyclonal
Unconjugated
FERMT2
Fermitin family homolog 2, FERMT2, KIND2, Kindlin 2, mig 2, MIG2, PLEKHC1, UNC112, UNC112B
Rabbit
Antigen Affinity Chromatography
RUO
10979, 218952, 289992
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.36 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8CIB5, Q96AC1
Fermt2
Kindlin 2 Fusion Protein Ag2013
150 μL
Primary
Human, Mouse, Rat
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote