missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA1199 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21129-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The function of this protein remains unknown.Spezifikation
KIAA1199 | |
Polyclonal | |
Unconjugated | |
CEMIP | |
CEMIP, KIAA1199, Protein KIAA1199, TMEM2L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
308797, 57214, 80982 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BI06, Q8WUJ3 | |
Cemip | |
KIAA1199 Fusion Protein Ag15527 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |