Learn More
KIAA1024L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553087
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MKNNPLYGDL SLEEAMEERK KNPSWTIEEY DKHSLHTNLS GHLKENPNDL RFWLGDMYTP GFDTLLKKEE KQEKHSKFCR M Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.
KIAA1024L Polyclonal antibody specifically detects KIAA1024L in Human samples. It is validated for Immunohistochemistry.Spezifikation
KIAA1024L | |
Polyclonal | |
Unconjugated | |
KIAA1024L | |
KIAA1024 like; KIAA1024L; KIAA1024-like; Major intrinsically disordered NOTCH2-binding receptor 1-like; Membrane integral NOTCH2-associated receptor 2; MINAR2; UPF0258 protein KIAA1024-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100127206 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P59773 | |
KIAA1024L | |
Recombinant protein corresponding to Human KIAA1024L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only