Learn More
KIAA0196 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA566702
Additional Details : Gewicht : 0.01000kg
Beschreibung
Immunogen sequence: ELVSYVRKVLQ IIPESMFTSL LKIIKLQTHD IIEVPTRLDK DKLRDYAQLG PRYEVAKLTH AISIFTEGIL MMKTTLVGII KVDPKQLLED GIRK Highest antigen sequence identity to the following orthologs - mouse 99%, rat 100%.
This gene encodes a 134 kDa protein named strumpellin that is predicted to have multiple transmembrane domains and a spectrin-repeat-containing domain. This ubiquitously expressed gene has its highest expression in skeletal muscle. The protein is named for Strumpell disease; a form of hereditary spastic paraplegia (HSP). Spastic paraplegias are a diverse group of disorders in which the autosomal dominant forms are characterized by progressive, lower extremity spasticity caused by axonal degeneration in the terminal portions of the longest descending and ascending corticospinal tracts. More than 30 loci (SPG1-33) have been implicated in hereditary spastic paraplegia diseases.Spezifikation
KIAA0196 | |
Polyclonal | |
Unconjugated | |
WASHC5 | |
AL022848; C76463; E430025E21Rik; KIAA0196; mKIAA0196; RGD1564420; RIKEN cDNA E430025E21 gene; RTSC; RTSC1; similar to Hypothetical protein MGC31278; SPG8; Strumpellin; uncharacterized protein LOC362919; WASH complex subunit 5; WASH complex subunit strumpellin; WASHC5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9897 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q12768 | |
WASHC5 | |
Recombinant Human KIAA0196 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only