missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ki67/MKI67 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
342.00 CHF - 512.00 CHF
Spezifikation
| Antigen | Ki67/MKI67 |
|---|---|
| Anwendungen | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Wirtsspezies | Rabbit |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18204640
|
Novus Biologicals
NBP2-54657 |
100 μL |
512.00 CHF
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18672867
|
Novus Biologicals
NBP2-54657-25ul |
25 μL |
342.00 CHF
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Ki67/MKI67 Polyclonal antibody specifically detects Ki67/MKI67 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Spezifikation
| Ki67/MKI67 | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 4288 | |
| This KI67/MKI67 Antibody was developed against a Recombinant Protein corresponding to amino acids 16-159 from Human KI67/MKI67: DGPHFPLSLSTCLFGRGIECDIRIQLPVVSKQHCKIEIHEQEAILHNFSSTNPTQVNGSVIDEPVRLKHGDVITIIDRSFRYENESLQSGRKSTEFPRKIREQEPARRVSRSSFSSDPDEKAQDSKA | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
| MKI67 | |
| IgG | |
| Affinity Purified | |
| 359 kDa |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts