missing translation for 'onlineSavingsMsg'
Learn More

Keratin 32 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA568042

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 15873024

  • 602.00 CHF / 100 Mikroliter
Verfügbar ab: 29-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Keratin 32 Polyclonal antibody specifically detects Keratin 32 in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

Keratin 32
Polyclonal
Unconjugated
KRT32
HA2; hair keratin; Hair keratin, type I Ha2; hard keratin, type I, 2; HHA2; HKA2; K32; Ka26; keratin 32; keratin 32, type I; keratin, hair, acidic, 2; keratin, type I cuticular Ha2; Keratin-32; KRT32; KRTHA2; type I cuticular hair keratin; type I hair keratin KA26
Rabbit
affinity chromatography
RUO
3882
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q14532
KRT32
Synthesized peptide derived from human Keratin 32.
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote