missing translation for 'onlineSavingsMsg'
Learn More

KDELR3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Thermo Scientific PA560377

Additional Details : Gewicht : 0.02000kg

Artikelnummer. 15738211

  • 654.00 CHF / 100 Mikroliter
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Immunogen sequence: FYLYVTKGRS WDDSNADTGL RSYSSI Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 38%.

KDELR3 Polyclonal antibody specifically detects KDELR3 in Human samples. It is validated for Immunohistochemistry.
Spezifikation

Spezifikation

KDELR3
Polyclonal
Unconjugated
KDELR3
AI173274; ER lumen protein retaining receptor 3; ER lumen protein-retaining receptor 3; ERD23; ERD2L3; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 3; KDEL endoplasmic reticulum protein retention receptor 3; KDEL receptor 3; Kdelr3
Rabbit
Antigen affinity chromatography
RUO
11015
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O43731
KDELR3
Recombinant protein corresponding to Human KDELR3
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Research Use Only