Learn More
KCTD18 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA566125
Additional Details : Gewicht : 0.01000kg
Beschreibung
Immunogen sequence: YPYSLSDHLAN EMETYSLRSN IELKKALTDF CDSYGLVCNK PTVWVLHYLN TSGASCESRI IGVYATKTDG TDAIEK Highest antigen sequence identity to the following orthologs - mouse 99%, rat 97%.
KCTD18 Polyclonal antibody specifically detects KCTD18 in Human samples. It is validated for Immunocytochemistry.Spezifikation
KCTD18 | |
Polyclonal | |
Unconjugated | |
KCTD18 | |
4932411A20Rik; 6530404F10Rik; BTB/POZ domain-containing protein KCTD18; KCTD18; potassium channel tetramerisation domain containing 18; potassium channel tetramerization domain containing 18; RGD1564820; similar to potassium channel tetramerisation domain containing 18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
130535 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PI47 | |
KCTD18 | |
Recombinant Human KCTD18 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only