missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550434
Additional Details : Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total KCNK13 protein.
KCNK13 Polyclonal antibody specifically detects KCNK13 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KCNK13 | |
Polyclonal | |
Unconjugated | |
KCNK13 | |
BB085247; F730021E22Rik; Gm1570; Gm1685; K2p13.1; K2P13.1 potassium channel; KCNK13; Potassium channel subfamily K member 13; potassium channel, subfamily K, member 13; potassium channel, two pore domain subfamily K, member 13; potassium two pore domain channel subfamily K member 13; prdx1; Tandem pore domain halothane-inhibited potassium channel 1; tandem pore domain potassium channel THIK-1; THIK1; THIK-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
217826, 56659 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8R1P5, Q9HB14 | |
KCNK13 | |
Fusion protein corresponding to a region derived from internal residues of human potassium channel, subfamily K, member 13 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |