missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNJ1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20953-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. It is activated by internal ATP and probably plays an important role in potassium homeostasis. The encoded protein has a greater tendency to allow potassium to flow into a cell rather than out of a cell. Mutations in this gene have been associated with antenatal Bartter syndrome, which is characterized by salt wasting, hypokalemic alkalosis, hypercalciuria, and low blood pressure. Multiple transcript variants encoding different isoforms have been found for this gene.Spezifikation
KCNJ1 | |
Polyclonal | |
Unconjugated | |
KCNJ1 | |
KCNJ1, KIR1.1, ROMK, ROMK1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
24521, 3758, 56379 | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88335, P35560, P48048 | |
KCNJ1 | |
KCNJ1 Fusion Protein Ag15100 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |