Learn More
KCC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA564474
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ILKQMHLTKN EREREIQSIT DESRGSIRRK NPANTRLRLN VPEETAGDSE EKPEEEVQLI HDQSAPSC Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 81%.
Mediates electroneutral potassium-chloride cotransport in mature neurons. Transport occurs under isotonic conditions, but is activated 20-fold by cell swelling. Important for Cl(-) homeostasis in neurons.Spezifikation
KCC2 | |
Polyclonal | |
Unconjugated | |
SLC12A5 | |
EIEE34; EIG14; Electroneutral potassium-chloride cotransporter 2; erythroid K-Cl cotransporter 2; furosemide-sensitive K-Cl cotransporter; hKCC2; KCC 2; Kcc2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; K-Cl cotransporter 2; KIAA1176; mKCC2; mKIAA1176; neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; S12A5; SLC12A5; solute carrier family 12 (potassium/chloride transporter), member 5; solute carrier family 12 (potassium-chloride transporter), member 5; solute carrier family 12 member 5; solute carrier family 12, (potassium-chloride transporter) member 5; solute carrier family 12, member 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57468 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H2X9 | |
SLC12A5 | |
Recombinant protein corresponding to Human KCC2 | |
100 μL | |
Primary | |
Human | |
Antibody | |