Learn More
KCC2 Monoclonal Antibody (S1-12), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA527610
Detalles adicionales : Gewicht : 0.01000kg
Descripción
1 µg/mL of MA5-27610 was sufficient for detection of KCC2 in 10 µg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 140kDa.
Mediates electroneutral potassium-chloride cotransport in mature neurons. Transport occurs under isotonic conditions, but is activated 20-fold by cell swelling. Important for Cl(-) homeostasis in neurons.Especificaciones
KCC2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
Q63633, Q91V14, Q9H2X9 | |
SLC12A5 | |
Fusion protein amino acids 932-1043 corresponding to rat KCC2 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
S1-12 | |
Unconjugated | |
SLC12A5 | |
EIEE34; EIG14; Electroneutral potassium-chloride cotransporter 2; erythroid K-Cl cotransporter 2; furosemide-sensitive K-Cl cotransporter; hKCC2; KCC 2; Kcc2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; K-Cl cotransporter 2; KIAA1176; mKCC2; mKIAA1176; neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; S12A5; SLC12A5; solute carrier family 12 (potassium/chloride transporter), member 5; solute carrier family 12 (potassium-chloride transporter), member 5; solute carrier family 12 member 5; solute carrier family 12, (potassium-chloride transporter) member 5; solute carrier family 12, member 5 | |
Mouse | |
Protein G | |
RUO | |
171373, 57138, 57468 | |
-20°C | |
Liquid |