missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kallikrein 4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5111343
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: TIRSISIASQ CPTAGNSCLV SGWGLLANGR MPTVLQCVNV SVVSEEVCSK LYDPLYH Highest antigen sequence indentity to the following orthologs: Mouse - 68%, Rat - 68%.
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel Kallikrein gene Prostase/KLK-L1 (also known as KLK4) is expressed and is up-regulated by androgens and progestins. KLK-L1 may be involved in the pathogenesis and/or progression of prostate, breast, and possibly other malignancies.
Spezifikation
| Kallikrein 4 | |
| Polyclonal | |
| Unconjugated | |
| KLK4 | |
| AI2A1; androgen-regulated message 1; ARM1; EMSP; Emsp1; enamel matrix serine protease 1; Enamel matrix serine proteinase 1; ESMP1; kallikrein; kallikrein 4 (prostase, enamel matrix, prostate); kallikrein related peptidase 4; kallikrein related-peptidase 4 (prostase, enamel matrix, prostate); kallikrein-4; Kallikrein-like protein 1; kallikrein-related peptidase 4; Klk4; KLK-4; KLKL1; KLK-L1; Prostase; protease, serine 17 (enamel matrix, prostate); Prss17; PSTS; serine protease 17 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9622 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y5K2 | |
| KLK4 | |
| Recombinant Protein corresonding to Human Kallikrein 4. Recombinant protein control fragment (Product #RP-101456). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur