missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JSRP1 Monoclonal Antibody (6A9), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA524513
Additional Details : Gewicht : 0.01000kg
Beschreibung
Peptide Sequence: MSMTTRAWEE LDGGLGSCQA LEDHSALAET QEDRASATPR LADSGSVPHD SQVAEGPSVD TRPKKMEKEP AARGTPGTGK ERLKAGASPR SVPARKKAQT
JSRP1 Monoclonal antibody specifically detects JSRP1 in Human samples. It is validated for ELISA, Western Blot.Spezifikation
JSRP1 | |
Monoclonal | |
Unconjugated | |
JSRP1 | |
2300003C06Rik; 2310032K21Rik; homolog of mouse skeletal muscle sarcoplasmic reticulum protein JP-45; JP45; JP-45; Jsrp1; Junctional sarcoplasmic reticulum protein 1; Junctional SR; junctional-face membrane protein of 45 kDa homolog; skeletal muscle sarcoplasmic reticulum protein JP-45 | |
Mouse | |
Protein A | |
RUO | |
126306 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
6A9 | |
PBS with no preservative; pH 7.4 | |
Q96MG2 | |
JSRP1 | |
JSRP1 (NP_653217.1, 1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |