missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JDP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550994
Additional Details : Gewicht : 0.01000kg
Beschreibung
JDP2 Polyclonal Antibody detects endogenous levels of JDP2 protein.
JDP2 Polyclonal antibody specifically detects JDP2 in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot.Spezifikation
JDP2 | |
Polyclonal | |
Unconjugated | |
JDP2 | |
c-Jun dimerization protein 2; Jdp2; Jdp-2; Jun dimerization protein 2; Jundm2; Jundp2; progesterone receptor co-activator; TIF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116674, 122953, 81703 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P97875, Q78E65, Q8WYK2 | |
JDP2 | |
Synthesized peptide derived from human JDP2 around the non-phosphorylation site of T148. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |