missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAMP Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP2-36446F
Additional Details : Gewicht : 0.00970kg
Beschreibung
JAMP Polyclonal specifically detects JAMP in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Spezifikation
JAMP | |
Polyclonal | |
1.0 mg/mL | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
C14orf100, chromosome 14 open reading frame 100, HSPC213, HSPC327, JAMPCDA06, JNK1/MAPK8-associated membrane protein, JNK1-associated membrane protein, JNK-associated membrane protein, Jun N-terminal kinase 1-associated membrane protein, Medulloblastoma antigen MU-MB-50.4 | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
51528 | |
Human | |
Purified |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
7A9-3A3 | |
FITC | |
PBS with 0.05% Sodium Azide | |
JKAMP | |
A synthetic peptide made to an internal portion of the human JAMP protein (between amino acids 20-150) [UniProt Q9P055] | |
0.1 mL | |
Primary | |
Cas9 protein from Streptococcus pyogene serotype M1. | |
Store at 4C in the dark. | |
IgG |
For Research Use Only