Learn More
JAKMIP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554625
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: IQARTVLTLA EKWLQQIEET EAALQRKMVD LESEKELFSK QKGYLDEELD YRKQALDQAN KHILELEAML YDALQQEAGA KVAELLSEEE REKLKVAVEQ WKRQVMSELR ERDAQILRER MELLQLAQQR IKELEERIEA QKRQIKELEE Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 97%.
JAKMIP3 gene ontology annotations related to this gene include kinase binding; microtubule binding.Spezifikation
JAKMIP3 | |
Polyclonal | |
Unconjugated | |
JAKMIP3 | |
6330417G02Rik; AI593226; bA140A10.5; C10orf14; C10orf39; JAKMIP3; JAMIP3; Janus kinase and microtubule interacting protein 3; Janus kinase and microtubule interacting protein 3; janus kinase and microtubule-interacting protein 3; janus kinase and microtubule-interacting protein 3; Kiaa4091; mKIAA4091; NECC2; neuroendocrine long coiled-coil 2; neuroendocrine long coiled-coil protein 2; RGD1307177 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
282973 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VZ66 | |
JAKMIP3 | |
Recombinant protein corresponding to Human JAKMIP3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only