missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGBL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA572575
Additional Details : Gewicht : 0.01000kg
Beschreibung
The protein encoded by this gene is a component of the F-type ATPase found in the mitochondrial matrix. F-type ATPases are composed of a catalytic core and a membrane proton channel. The encoded protein appears to be part of the connector linking these two components and may be involved in transmission of conformational changes or proton conductance.Spezifikation
ITGBL1 | |
Polyclonal | |
Unconjugated | |
ITGBL1 | |
B930011D01Rik; integrin beta like 1; integrin beta-like protein 1; integrin subunit beta like 1; integrin, beta-like 1; integrin, beta-like 1 (with EGF-like repeat domains); ITGBL1; OSCP; Osteoblast-specific cysteine-rich protein; ten integrin EGF-like repeat domain-containing protein; TIED; with EGF-like repeat domains | |
Rabbit | |
Protein A | |
RUO | |
9358 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
O95965 | |
ITGBL1 | |
KLH conjugated synthetic peptide between 334-360 amino acids from the Central region of human ITGBL1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |